accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSH

CAS RN 371-42-6

IUPAC Name: 4-fluorobenzene-1-thiol
Synonyms: 4-fluorobenzenethiol
Molecular Weight (g/mol): 128.16
Molecular Formula: C6H5FS
InChi Key: OKIHXNKYYGUVTE-UHFFFAOYSA-N
SMILES: FC1=CC=C(S)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)