CAS RN 37115-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ alpha-Hydroxyalprazolam Solution, 1.0 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This certified Snap-N-Spike™ solution standard is suitable for use in LC/MS or GC/MS applications such as forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.