accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiBFFFFFFFFFFFFFFFFFFFFCH3OH3C

CAS RN 371162-53-7

IUPAC Name: lithium(1+) ethoxyethane tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Synonyms: lithium tetrakis(pentafluorophenyl)borate
Molecular Weight (g/mol): 760.11
Molecular Formula: C28H10BF20LiO
InChi Key: KPLZKJQZPFREPG-UHFFFAOYSA-N
SMILES: [Li+].CCOCC.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results