accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3715-29-5

IUPAC Name: sodium 3-methyl-2-oxobutanoate
Synonyms: α-ketoisovaleric acid
Molecular Weight (g/mol): 138.1
Molecular Formula: C5H7NaO3
InChi Key: WIQBZDCJCRFGKA-UHFFFAOYSA-M
SMILES: [Na+].CC(C)C(=O)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results