accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NO

CAS RN 3718-65-8

IUPAC Name: 3,5-dimethylpyridin-1-ium-1-olate
Synonyms: 3,5-dimethylpyridine-N-oxide
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: SSTLCMOZTCLOLQ-UHFFFAOYSA-N
SMILES: CC1=C[N+]([O-])=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results