accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHF

CAS RN 372-20-3

IUPAC Name: 3-fluorophenol
Synonyms: M-fluorophenol
Molecular Weight (g/mol): 112.1
Molecular Formula: C6H5FO
InChi Key: SJTBRFHBXDZMPS-UHFFFAOYSA-N
SMILES: OC1=CC=CC(F)=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (4)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)