accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNClF

CAS RN 372183-70-5

IUPAC Name: 2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
Synonyms: 2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine
Molecular Weight (g/mol): 250.7
Molecular Formula: C13H12ClFN2
InChi Key: VALSYGYSZKIHMQ-UHFFFAOYSA-N
SMILES: CC1=CC(CCC2=CC=C(F)C=C2)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results