CAS RN 3722-51-8
CAS RN 3722-51-8
IUPAC Name:
3-hydroxy-9H-xanthen-9-one
Synonyms:
3-hydroxyxanthen-9-one
Molecular Weight (g/mol):
212.2
Molecular Formula:
C13H8O3
InChi Key:
XCJHDJAODLKGLG-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C(OC3=CC=CC=C3C2=O)=C1
1
–
1
of
1
results
1
3-Hydroxyxanthen-9-one 98.0+%, TCI America™
CAS: 3722-51-8 Molecular Formula: C13H8O3 Molecular Weight (g/mol): 212.204 MDL Number: MFCD00604748 InChI Key: XCJHDJAODLKGLG-UHFFFAOYSA-N Synonym: sieber linker,3-hydroxy-9h-xanthen-9-one,3-hydroxy-xanthen-9-one,3-hydroxyxanthone,9h-xanthen-9-one, 3-hydroxy,sieber linker;,xanthen-9-one, 3-hydroxy,pubchem12819,acmc-209irq,3-hydroxy-9-xanthenone PubChem CID: 5376013 IUPAC Name: 3-hydroxyxanthen-9-one SMILES: C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O