accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKKKOHSOOOHOSOOOOOSOOO

CAS RN 37222-66-5

IUPAC Name: tetrapotassium oxidanesulfonoperoxoate hydrogen sulfate sulfate
Synonyms: tetrapotassium sulfodioxidanide hydrogen sulfate sulfate
Molecular Weight (g/mol): 462.58
Molecular Formula: H2K4O13S3
InChi Key: TZTINBNQGPHMBC-UHFFFAOYSA-J
SMILES: [K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.OS(=O)(=O)O[O-]
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)