accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON

CAS RN 3724-19-4

IUPAC Name: 3-(pyridin-3-yl)propanoic acid
Synonyms: 3-(pyridin-3-yl)propanoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: WDGXIUUWINKTGP-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)