CAS RN 37271-17-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sennoside D, ≥98% (HPLC), MilliporeSigma™ Supelco™
Sennoside D is an active constituent of senna which is generally used as a laxative. It belongs to anthranoid family, and is procured from dried leaflets and pods of senna plant.