accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3728-43-6

IUPAC Name: trimethyl(4-methylphenyl)silane
Synonyms: trimethyl(4-methylphenyl)silane
Molecular Weight (g/mol): 164.32
Molecular Formula: C10H16Si
InChi Key: QGHURGPPCGMAMZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results