accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3C

CAS RN 3728-54-9

IUPAC Name: 1-ethyl-2-methylcyclohexane
Synonyms: 1-ethyl-2-methylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: XARGIVYWQPXRTC-UHFFFAOYNA-N
SMILES: CCC1CCCCC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results