accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NFFF

CAS RN 373-88-6

IUPAC Name: hydrogen 2,2,2-trifluoroethan-1-amine chloride
Synonyms: hydrogen 2,2,2-trifluoroethylamine chloride
Molecular Weight (g/mol): 135.51
Molecular Formula: C2H5ClF3N
InChi Key: ZTUJDPKOHPKRMO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)