accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3730-60-7

IUPAC Name: 2,5-dimethylhexan-2-ol
Synonyms: 2-hexanol, 2,5-dimethyl-, (S)-
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: JPUIYNHIEXIFMV-UHFFFAOYSA-N
SMILES: CC(C)CCC(C)(C)O
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)