accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBrFClH2C

CAS RN 374-25-4

IUPAC Name: 4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
Synonyms: 4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
Molecular Weight (g/mol): 223.42
Molecular Formula: C4H3BrClF3
InChi Key: VULPFOSLGWWARI-UHFFFAOYNA-N
SMILES: FC(F)(Br)C(F)(Cl)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)