CAS RN 3741-00-2
IUPAC Name:
pentylcyclopentane
Synonyms:
cyclopentane, pentyl-
Molecular Weight (g/mol):
140.27
Molecular Formula:
C10H20
InChi Key:
HPQURZRDYMUHJI-UHFFFAOYSA-N
SMILES:
CCCCCC1CCCC1
1
–
1
of
1
results
1
Pentylcyclopentane 99.0+%, TCI America™
CAS: 3741-00-2 Molecular Formula: C10H20 Molecular Weight (g/mol): 140.27 MDL Number: MFCD00039451 InChI Key: HPQURZRDYMUHJI-UHFFFAOYSA-N Synonym: Amylcyclopentane PubChem CID: 19540 IUPAC Name: pentylcyclopentane SMILES: CCCCCC1CCCC1