accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrOOCH2Br

CAS RN 3741-77-3

IUPAC Name: 2,4,6-tribromophenyl prop-2-enoate
Synonyms: 2,4,6-tribromophenyl prop-2-enoate
Molecular Weight (g/mol): 384.85
Molecular Formula: C9H5Br3O2
InChi Key: CNLVUQQHXLTOTC-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(OC(=O)C=C)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results