accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOO

CAS RN 37418-88-5

IUPAC Name: 4-hydroxy-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-hydroxy-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 164.12
Molecular Formula: C8H4O4
InChi Key: CCTOEAMRIIXGDJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)OC2=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)