accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37441-29-5

IUPAC Name: 5-amino-2,4,6-triiodobenzene-1,3-dicarbonyl dichloride
Synonyms: 5-amino-2,4,6-triiodobenzene-1,3-dicarbonyl dichloride
Molecular Weight (g/mol): 595.72
Molecular Formula: C8H2Cl2I3NO2
InChi Key: FBJVWRITWDYUAC-UHFFFAOYSA-N
SMILES: NC1=C(I)C(C(Cl)=O)=C(I)C(C(Cl)=O)=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results