accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFCl

CAS RN 374633-32-6

IUPAC Name: 2-chloro-3-fluoro-6-methylpyridine
Synonyms: 2-chloro-3-fluoro-6-methylpyridine
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: BGDYLOGCMJAUHR-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results