accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3747-74-8

IUPAC Name: hydrogen 2-(chloromethyl)quinoline chloride
Synonyms: hydrogen 2-(chloromethyl)quinoline chloride
Molecular Weight (g/mol): 214.09
Molecular Formula: C10H9Cl2N
InChi Key: WDETYCRYUBGKCE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCC1=CC=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)