accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2CCH3H2C

CAS RN 3748-13-8

IUPAC Name: 1,3-bis(prop-1-en-2-yl)benzene
Synonyms: 1,3-bis(prop-1-en-2-yl)benzene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: IBVPVTPPYGGAEL-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC(=CC=C1)C(C)=C
Percent Purity 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results