CAS RN 374922-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SIRT1 Activator II
The SIRT1 Activator II, also referenced under CAS 374922-43-7, modulates the biological activity of SIRT1. This small molecule/inhibitor is primarily used for Biochemicals applications.