accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37493-70-2

IUPAC Name: undecan-5-ol
Synonyms: undecan-5-ol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: PCFOZHPCKQPZCN-UHFFFAOYNA-N
SMILES: CCCCCCC(O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results