accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFF

CAS RN 375-61-1

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5-undecafluoropentane
Synonyms: 1,1,1,2,2,3,3,4,4,5,5-undecafluoropentane
Molecular Weight (g/mol): 270.05
Molecular Formula: C5HF11
InChi Key: WXFBZGUXZMEPIR-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results