accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOFFFFFFFFFFFFF

CAS RN 375-82-6

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptan-1-ol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptan-1-ol
Molecular Weight (g/mol): 350.08
Molecular Formula: C7H3F13O
InChi Key: STLNAVFVCIRZLL-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)