accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3HOOFFFFFFFFFFFFF

CAS RN 375-85-9

IUPAC Name: tridecafluoroheptanoic acid amine
Synonyms: perfluoroheptanoic acid amine
Molecular Weight (g/mol): 381.09
Molecular Formula: C7H4F13NO2
InChi Key: JVZREVRTMWNFME-UHFFFAOYSA-N
SMILES: N.OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)