accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CHNH3CH3CCH3

CAS RN 37500-95-1

IUPAC Name: 3,6-di-tert-butyl-9H-carbazole
Synonyms: 3,6-di-tert-butyl-9H-carbazole
Molecular Weight (g/mol): 279.43
Molecular Formula: C20H25N
InChi Key: OYFFSPILVQLRQA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results