accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClO

CAS RN 37517-81-0

IUPAC Name: methyl 3-chloro-3-oxopropanoate
Synonyms: methyl 3-chloro-3-oxopropanoate
Molecular Weight (g/mol): 136.53
Molecular Formula: C4H5ClO3
InChi Key: UTBCRHAMJFMIIR-UHFFFAOYSA-N
SMILES: COC(=O)CC(Cl)=O
Boiling Point 
  • (4)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (7)
  • (4)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (7)