accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2

CAS RN 37529-27-4

IUPAC Name: 4-heptylaniline
Synonyms: 4-heptylaniline
Molecular Weight (g/mol): 191.32
Molecular Formula: C13H21N
InChi Key: BNEWZYZRLNNWNR-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)