accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3753-05-7

HOOOOOHO

CAS RN 3753-05-7

IUPAC Name: 4-[2-(4-carboxyphenoxy)ethoxy]benzoic acid
Synonyms: 4-[2-(4-carboxyphenoxy)ethoxy]benzoic acid
Molecular Weight (g/mol): 302.28
Molecular Formula: C16H14O6
InChi Key: VAXBLYWAVAIJJJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OCCOC2=CC=C(C=C2)C(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results