accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37535-52-7

IUPAC Name: 2-amino-3-(pyridin-2-yl)propanoic acid
Synonyms: 2-amino-3-(pyridin-2-yl)propanoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: PDRJLZDUOULRHE-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)