CAS RN 37558-29-5
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylate
Synonyms:
2,5-dioxopyrrolidin-1-yl 1-hydroxy-2,2,5,5-tetramethylpyrrole-3-carboxylate
Molecular Weight (g/mol):
282.3
Molecular Formula:
C13H18N2O5
InChi Key:
VDSVTMQVXNIMMJ-UHFFFAOYSA-N
SMILES:
CC1(C)C=C(C(=O)ON2C(=O)CCC2=O)C(C)(C)N1O
1
–
1
of
1
results
1
Promotions Available
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid N-hydroxysuccinimide ester, 98+%
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid N-hydroxysuccinimide ester, 98+%
CAS: 37558-29-5 Molecular Formula: C13H17N2O5 MDL Number: MFCD00066369