accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3O

CAS RN 37593-02-5

IUPAC Name: 1-(4-pentylphenyl)ethan-1-one
Synonyms: 1-(4-pentylphenyl)ethanone
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: KBKGPMDADJLBEM-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results