accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFFFFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 376-06-7

IUPAC Name: heptacosafluorotetradecanoic acid
Synonyms: heptacosafluorotetradecanoic acid
Molecular Weight (g/mol): 714.12
Molecular Formula: C14HF27O2
InChi Key: RUDINRUXCKIXAJ-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)