accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376-27-2

CH3OOFFFFFFFFFFFFFFF

CAS RN 376-27-2

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
Molecular Weight (g/mol): 428.1
Molecular Formula: C9H3F15O2
InChi Key: XOCNYZFAMHDXJK-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results