accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3762-25-2

IUPAC Name: diethyl [(4-methylphenyl)methyl]phosphonate
Synonyms: diethyl (4-methylphenyl)methylphosphonate
Molecular Weight (g/mol): 242.26
Molecular Formula: C12H19O3P
InChi Key: QKGBKPZAXXBLJE-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(C)C=C1)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)