accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37632-18-1

IUPAC Name: (4-methoxyphenyl)phosphonoyl dichloride
Synonyms: 4-methoxyphenylphosphonoyl dichloride
Molecular Weight (g/mol): 225
Molecular Formula: C7H7Cl2O2P
InChi Key: LOFIFCKKPQFWSH-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P(Cl)(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results