accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNClN

CAS RN 3764-01-0

IUPAC Name: 2,4,6-trichloropyrimidine
Synonyms: pyrimidine, 2,4,6-trichloro-
Molecular Weight (g/mol): 183.42
Molecular Formula: C4HCl3N2
InChi Key: DPVIABCMTHHTGB-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NC(Cl)=N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)