accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376581-24-7

IUPAC Name: (quinolin-6-yl)boronic acid
Synonyms: quinolin-6-ylboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: JLOLSBLXNMVKGY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CN=C2C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results