accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376584-62-2

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-isoindol-1-one
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroisoindol-1-one
Molecular Weight (g/mol): 259.11
Molecular Formula: C14H18BNO3
InChi Key: CLACMCLRELMFLJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C(=O)NC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results