accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 376591-04-7

IUPAC Name: 1-bromononadeca-4,6-diyne
Synonyms: 1-bromononadeca-4,6-diyne
Molecular Weight (g/mol): 339.36
Molecular Formula: C19H31Br
InChi Key: BCWJEWJRRDCQQU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#CC#CCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results