accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONH2OH

CAS RN 376592-93-7

IUPAC Name: 3'-amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3'-amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 229.24
Molecular Formula: C13H11NO3
InChi Key: ZXLYSSHNDUXXIN-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1O)C1=CC(=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results