accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376637-85-3

IUPAC Name: 1-(3-phenoxyphenyl)methanamine hydrochloride
Synonyms: 1-(3-phenoxyphenyl)methanamine hydrochloride
Molecular Weight (g/mol): 235.71
Molecular Formula: C13H14ClNO
InChi Key: WMFHUUKYIUOHRA-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC(OC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results