accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37674-72-9

IUPAC Name: 6-chloro-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 6-chloro-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 182.6
Molecular Formula: C9H7ClO2
InChi Key: LLTDYHFVIVSQPJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OCCC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results