accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHN

CAS RN 3770-50-1

IUPAC Name: ethyl 1H-indole-2-carboxylate
Synonyms: ethyl 1H-indole-2-carboxylate
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: QQXQAEWRSVZPJM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)