accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 377085-56-8

IUPAC Name: 4-{[(pentyloxy)carbonyl]oxy}benzoic acid
Synonyms: 4-{[(pentyloxy)carbonyl]oxy}benzoic acid
Molecular Weight (g/mol): 252.27
Molecular Formula: C13H16O5
InChi Key: RMIZZENBDILCRU-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)OC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results