accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3772-13-2

IUPAC Name: 2,2-dimethylthiirane
Synonyms: 2,2-dimethylthiirane
Molecular Weight (g/mol): 88.17
Molecular Formula: C4H8S
InChi Key: HGJOFJDIHKHKAU-UHFFFAOYSA-N
SMILES: CC1(C)CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results