accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37721-75-8

IUPAC Name: 2,3,5,6-tetrabromo-4-methylphenol
Synonyms: 2,3,5,6-tetrabromo-4-methylphenol
Molecular Weight (g/mol): 423.72
Molecular Formula: C7H4Br4O
InChi Key: OMVMKSWFUQZIFD-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(Br)=C(O)C(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results